Assay Details
Target Gene Details
Entrez Gene ID: | 117248 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 15 |
Gene Aliases: |
GALNACT15, GALNT13, GALNT7, GALNTL2, PIH5, pp-GalNAc-T15 |
Location: |
Chr.3:16174322-16245190 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT15 | NM_001319051.1 | 1 | 454 | NP_001305980.1 |
| NM_054110.4 | 1 | 99 | NP_473451.3 | |
| AK094780.1 | ||||
| AK225673.1 | 1 | 124 | ||
| AK312425.1 | 1 | 100 | ||
| AL831925.1 | 1 | 127 | CAD38585.1 | |
| AL832736.1 | 1 | 69 | ||
| AY358443.1 | 1 | 85 | AAQ88808.1 | |
| BC014789.1 | 1 | 49 | AAH14789.1 | |
| DA279091.1 | 1 | 454 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv589814 | Chr.3:16165768 - 16193535 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map