Assay Details
Target Gene Details
Entrez Gene ID: | 108 |
Gene Name: | adenylate cyclase 2 |
Gene Aliases: |
AC2, HBAC2 |
Location: |
Chr.5:7396230-7830081 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCY2 | NM_020546.2 | 1 | 192 | NP_065433.2 |
| XM_011513942.2 | 1 | 139 | XP_011512244.1 | |
| AF410885.1 | 1 | 151 | AAP97285.1 | |
| AK127036.1 | 1 | 140 | ||
| AK304041.1 | ||||
| BC111743.1 | 1 | 130 | AAI11744.1 | |
| BE677757.1 | 1 | 190 | ||
| DB481853.2 | 1 | 192 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830198 | Chr.5:7340105 - 7511680 on Build GRCh38 | Gain |
|
| nsv596996 | Chr.5:7359744 - 7447412 on Build GRCh38 | Loss |
|
| nsv596997 | Chr.5:7395937 - 7396713 on Build GRCh38 | Gain |
|
| nsv596998 | Chr.5:7396313 - 7398433 on Build GRCh38 | Loss |
|
| esv23386 | Chr.5:7395910 - 7396832 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map