Assay Details
Target Gene Details
Entrez Gene ID: | 11251 |
Gene Name: | prostaglandin D2 receptor 2 |
Gene Aliases: |
CD294, CRTH2, DL1R, DP2, GPR44 |
Location: |
Chr.11:60850925-60855971 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGDR2 | NM_004778.2 | 2 | 2362 | NP_004769.2 |
| AB008535.1 | 2 | 2362 | BAA74518.1 | |
| AF144308.1 | 2 | 2341 | AAD34539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952456 | Chr.11:60847528 - 60852527 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map