Assay Details
Target Gene Details
Entrez Gene ID: | 1147 |
Gene Name: | conserved helix-loop-helix ubiquitous kinase |
Gene Aliases: |
IKBKA, IKK-alpha, IKK1, IKKA, NFKBIKA, TCF16 |
Location: |
Chr.10:100188298-100229610 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHUK | NM_001278.4 | 21 | 2817 | NP_001269.3 |
| NM_001320928.1 | 21 | 2800 | NP_001307857.1 | |
| AB209028.1 | 20 | 5100 | BAD92265.1 | |
| AF080157.1 | 21 | 2834 | AAD08996.1 | |
| AL133012.1 | 3 | 4116 | ||
| BC092514.1 | 21 | 2755 | AAH92514.1 |
More Information
Set Membership: |
|

Genomic Map