Assay Details
Target Gene Details
Entrez Gene ID: | 6301 |
Gene Name: | seryl-tRNA synthetase |
Gene Aliases: |
SERRS, SERS |
Location: |
Chr.1:109213893-109238182 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SARS | NM_006513.3 | 11 | 1643 | NP_006504.2 |
| XM_006710813.2 | 12 | 1709 | XP_006710876.1 | |
| AK093718.1 | 2 | 1997 | ||
| AK222677.1 | 11 | 1604 | BAD96397.1 | |
| AK312771.1 | ||||
| BC000716.1 | 11 | 1552 | AAH00716.1 | |
| BC009390.2 | 11 | 1590 | AAH09390.1 | |
| BC111398.1 | 11 | 1613 | ||
| BM666450.1 | 1 | 314 | ||
| D49914.1 | 11 | 1546 | BAA95602.1 | |
| X91257.1 | 11 | 1618 | CAA62635.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012267 | Chr.1:109168205 - 109279887 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map