Assay Details
Target Gene Details
Entrez Gene ID: | 130589 |
Gene Name: | galactose mutarotase |
Gene Aliases: |
BLOCK25, GLAT, HEL-S-63p, IBD1 |
Location: |
Chr.2:38665910-38734767 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALM | NM_138801.2 | 1 | 242 | NP_620156.1 |
| XM_011532540.1 | 1 | 242 | XP_011530842.1 | |
| AK291489.1 | ||||
| BC014916.1 | AAH14916.1 | |||
| BC019263.1 | AAH19263.1 | |||
| DA984992.1 | 1 | 242 | ||
| EU794611.1 | ||||
| U11036.1 | AAA67652.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3590419 | Chr.2:38660017 - 38726534 on Build GRCh38 | Loss |
|
| nsv581485 | Chr.2:38612181 - 38666518 on Build GRCh38 | Gain |
|
| nsv9646 | Chr.2:38131169 - 38855047 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map