Assay Details
Target Gene Details
Entrez Gene ID: | 1240 |
Gene Name: | chemerin chemokine-like receptor 1 |
Gene Aliases: |
CHEMERINR, ChemR23, DEZ, RVER1 |
Location: |
Chr.12:108288044-108339348 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMKLR1 | NM_001142343.1 | 4 | 1350 | NP_001135815.1 |
| NM_001142344.1 | 3 | 1137 | NP_001135816.1 | |
| NM_001142345.1 | 3 | 964 | NP_001135817.1 | |
| NM_004072.2 | 3 | 1274 | NP_004063.1 | |
| XM_017018820.1 | 2 | 1092 | XP_016874309.1 | |
| AB112777.1 | 1 | 767 | ||
| AK291800.1 | 3 | 1131 | ||
| AY497547.1 | 1 | 767 | AAR90850.1 | |
| BC106927.2 | 4 | 1164 | AAI06928.1 | |
| BC106928.1 | 4 | 1164 | AAI06929.1 | |
| BT019556.1 | 1 | 767 | AAV38363.1 | |
| BT019557.1 | 1 | 767 | AAV38364.1 | |
| U79526.1 | 3 | 1127 | AAC51258.1 | |
| U79527.1 | 3 | 1280 | AAC51259.1 |
More Information
Set Membership: |
|

Genomic Map