Assay Details
Target Gene Details
Entrez Gene ID: | 55571 |
Gene Name: | CCR4-NOT transcription complex subunit 11 |
Gene Aliases: |
C2orf29, C40 |
Location: |
Chr.2:101252883-101270316 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT11 | NM_017546.4 | NP_060016.3 | ||
| AF103798.1 | AAF02418.1 | |||
| AK024221.1 | ||||
| BC018664.2 | AAH18664.2 | |||
| BC064421.1 | AAH64421.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34123 | Chr.2:101008454 - 101391851 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map