Assay Details
Target Gene Details
Entrez Gene ID: | 30817 |
Gene Name: | adhesion G protein-coupled receptor E2 |
Gene Aliases: |
CD312, EMR2, VBU |
Location: |
Chr.19:14732697-14778541 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRE2 | NM_001271052.1 | NP_001257981.1 | ||
| NM_013447.3 | NP_038475.2 | |||
| XM_011527948.2 | XP_011526250.1 | |||
| XM_011527949.2 | XP_011526251.1 | |||
| XM_011527951.2 | XP_011526253.1 | |||
| XM_011527952.2 | XP_011526254.1 | |||
| XM_011527953.2 | XP_011526255.1 | |||
| XM_011527954.2 | XP_011526256.1 | |||
| XM_011527955.2 | XP_011526257.1 | |||
| XM_017026726.1 | XP_016882215.1 | |||
| XM_017026727.1 | XP_016882216.1 | |||
| AF114491.1 | AAF21974.1 | |||
| AK024426.1 | BAB15716.1 | |||
| AK222891.1 | BAD96611.1 | |||
| AK291776.1 | ||||
| AK298690.1 | ||||
| AK298700.1 | ||||
| AK301458.1 | ||||
| BC034949.1 | AAH34949.1 | |||
| BC053933.1 | AAH53933.1 | |||
| BC127004.1 | ||||
| BC127005.1 | ||||
| CR936819.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953982 | Chr.19:14758889 - 14781488 on Build GRCh38 | Deletion |
|
| nsv509722 | Chr.19:14750795 - 14803000 on Build GRCh38 | Insertion |
|
| esv3893167 | Chr.19:14610447 - 14950404 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map