Assay Details
Target Gene Details
Entrez Gene ID: | 2539 |
Gene Name: | glucose-6-phosphate dehydrogenase |
Gene Aliases: |
G6PD1 |
Location: |
Chr.X:154531390-154547586 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| G6PD | NM_001042351.2 | NP_001035810.1 | ||
| AL560686.3 | ||||
| DC394789.1 |
Target Gene Details
Entrez Gene ID: | 8517 |
Gene Name: | inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma |
Gene Aliases: |
AMCBX1, FIP-3, FIP3, Fip3p, IKK-gamma, IKKAP1, IKKG, IMD33, IP, IP1, IP2, IPD2, NEMO, ZC2HC9 |
Location: |
Chr.X:154542240-154565046 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IKBKG | NM_001099856.4 | NP_001093326.2 | ||
| NM_001099857.2 | 1 | 178 | NP_001093327.1 | |
| NM_001145255.2 | 1 | 178 | NP_001138727.1 | |
| NM_001321396.1 | NP_001308325.1 | |||
| NM_001321397.1 | 1 | 178 | NP_001308326.1 | |
| AF074382.1 | AAC36330.1 | |||
| AK308366.1 | 1 | 178 | ||
| AY114157.1 | AAM44073.1 | |||
| BC012114.1 | AAH12114.1 | |||
| BM473416.1 | ||||
| DC410130.1 | 1 | 178 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss |
|
| nsv519042 | Chr.X:154082978 - 154601444 on Build GRCh38 | Gain |
|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
| nsv508816 | Chr.X:154492171 - 154591783 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map