Assay Details
Target Gene Details
Entrez Gene ID: | 9508 |
Gene Name: | ADAM metallopeptidase with thrombospondin type 1 motif 3 |
Gene Aliases: |
ADAMTS-4 |
Location: |
Chr.4:72280969-72568799 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTS3 | NM_014243.2 | 22 | 5348 | NP_055058.2 |
| XM_011532421.2 | 22 | 5380 | XP_011530723.1 | |
| XM_011532422.2 | 22 | 5879 | XP_011530724.1 | |
| AB002364.1 | 22 | 5300 | BAA20821.1 | |
| BC016451.1 | 5 | 2788 | AAH16451.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829968 | Chr.4:72147632 - 72286232 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map