Assay Details
Target Gene Details
Entrez Gene ID: | 10526 |
Gene Name: | importin 8 |
Gene Aliases: |
RANBP8 |
Location: |
Chr.12:30628981-30695995 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IPO8 | NM_001190995.1 | NP_001177924.1 | ||
| NM_006390.3 | NP_006381.2 | |||
| XM_017018691.1 | XP_016874180.1 | |||
| XM_017018692.1 | XP_016874181.1 | |||
| AB209602.1 | BAD92839.1 | |||
| AK302260.1 | ||||
| AU132276.3 | ||||
| U77494.1 | AAB67052.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832361 | Chr.12:30641096 - 30844307 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map