Assay Details
Target Gene Details
Entrez Gene ID: | 22877 |
Gene Name: | MLX interacting protein |
Gene Aliases: |
MIR, MONDOA, bHLHe36 |
Location: |
Chr.12:122078728-122147347 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLXIP | NM_014938.5 | NP_055753.3 | ||
| XM_006719290.3 | XP_006719353.1 | |||
| XM_006719291.3 | XP_006719354.1 | |||
| XM_006719292.3 | XP_006719355.1 | |||
| XM_006719293.3 | XP_006719356.1 | |||
| AF312918.1 | AAG34121.1 | |||
| BC028309.1 | AAH28309.1 | |||
| BC151841.1 | ||||
| BM461435.1 |
More Information
Set Membership: |
|

Genomic Map