Assay Details
Target Gene Details
Entrez Gene ID: | 1407 |
Gene Name: | cryptochrome circadian clock 1 |
Gene Aliases: |
PHLL1 |
Location: |
Chr.12:106991364-107093872 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRY1 | NM_004075.4 | 13 | 3213 | NP_004066.1 |
| XM_017018832.1 | 14 | 3348 | XP_016874321.1 | |
| AK098615.1 | 8 | 1617 | BAC05354.1 | |
| AK125915.1 | 13 | 3755 | ||
| BC030519.1 | 13 | 2886 | AAH30519.1 | |
| D83702.1 | 13 | 2903 | BAA12068.1 |
More Information
Set Membership: |
|

Genomic Map