Assay Details
Target Gene Details
Entrez Gene ID: | 78999 |
Gene Name: | leucine rich repeat and fibronectin type III domain containing 4 |
Gene Aliases: |
FIGLER6, SALM3, SALM3. |
Location: |
Chr.11:66857087-66860475 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRFN4 | NM_024036.4 | NP_076941.2 | ||
| XM_005274239.3 | XP_005274296.1 | |||
| AB209005.1 | BAD92242.1 | |||
| BC000207.1 | AAH00207.2 | |||
| BC007718.2 | AAH07718.1 | |||
| BC014040.2 | AAH14040.2 | |||
| BC015581.2 | AAH15581.2 | |||
| BC027475.1 | AAH27475.2 | |||
| BC071866.1 | AAH71866.1 | |||
| BC094813.1 | AAH94813.1 |
Target Gene Details
Entrez Gene ID: | 5091 |
Gene Name: | pyruvate carboxylase |
Gene Aliases: |
PCB |
Location: |
Chr.11:66848522-66958418 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PC | NM_000920.3 | NP_000911.2 | ||
| NM_001040716.1 | NP_001035806.1 | |||
| NM_022172.2 | NP_071504.2 | |||
| XM_005274031.4 | XP_005274088.1 | |||
| XM_005274032.4 | XP_005274089.1 | |||
| XM_006718578.3 | XP_006718641.1 | |||
| XM_011545086.2 | XP_011543388.1 | |||
| XM_017017868.1 | XP_016873357.1 | |||
| XM_017017869.1 | XP_016873358.1 | |||
| XM_017017870.1 | XP_016873359.1 | |||
| XM_017017871.1 | XP_016873360.1 | |||
| XM_017017872.1 | XP_016873361.1 | |||
| BC011617.2 | AAH11617.1 | |||
| S72370.1 | AAB31500.1 | |||
| U04641.1 | AAA99537.1 | |||
| U30891.1 | AAA82937.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555226 | Chr.11:66812101 - 67077891 on Build GRCh38 | Gain |
|
| nsv951020 | Chr.11:66843130 - 66869629 on Build GRCh38 | Deletion |
|
| nsv508638 | Chr.11:66774182 - 66876163 on Build GRCh38 | Deletion |
|
| nsv468602 | Chr.11:66700998 - 66866926 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map