Assay Details
Target Gene Details
Entrez Gene ID: | 80018 |
Gene Name: | N(alpha)-acetyltransferase 25, NatB auxiliary subunit |
Gene Aliases: |
C12orf30, MDM20, NAP1 |
Location: |
Chr.12:112026689-112109022 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA25 | NM_024953.3 | 18 | 2239 | NP_079229.2 |
| XM_005253938.1 | 17 | 2085 | XP_005253995.1 | |
| XM_006719606.2 | 18 | 2899 | XP_006719669.1 | |
| XM_017019977.1 | 16 | 1955 | XP_016875466.1 | |
| AB054990.1 | 18 | 2181 | BAC80174.1 | |
| AK023151.1 | 15 | 1884 | BAB14432.1 | |
| AK292485.1 | 18 | 2212 | ||
| AK307494.1 | 19 | 2388 | ||
| AK309772.1 | 18 | 2194 | ||
| AL832394.1 | 2 | 307 | ||
| BC034357.1 | 18 | 2193 | ||
| BC113585.1 | 18 | 2239 | ||
| BC113587.1 | 18 | 2239 | ||
| BX641140.1 | 18 | 2194 | CAE46062.1 | |
| BX647450.1 | 18 | 2192 | ||
| BX647992.1 | 16 | 1969 |
More Information
Set Membership: |
|

Genomic Map