Assay Details
Target Gene Details
Entrez Gene ID: | 4359 |
Gene Name: | myelin protein zero |
Gene Aliases: |
CHM, CMT1, CMT1B, CMT2I, CMT2J, CMT4E, CMTDI3, CMTDID, DSS, HMSNIB, MPP, P0 |
Location: |
Chr.1:161303593-161309972 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPZ | NM_000530.7 | 6 | 1262 | NP_000521.2 |
| NM_001315491.1 | 6 | 1262 | NP_001302420.1 | |
| XM_017001321.1 | XP_016856810.1 | |||
| BF509916.1 | ||||
| CD172418.1 | 1 | 406 | ||
| CD515400.1 | 1 | 517 | ||
| CD515605.1 | 3 | 736 | ||
| D10537.1 | 6 | 1238 | BAA01395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517826 | Chr.1:161293590 - 161307811 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map