Assay Details
Target Gene Details
Entrez Gene ID: | 84958 |
Gene Name: | synaptotagmin like 1 |
Gene Aliases: |
JFC1, SLP1 |
Location: |
Chr.1:27341992-27353932 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYTL1 | NM_001193308.1 | 3 | 492 | NP_001180237.1 |
| NM_032872.2 | 3 | 492 | NP_116261.1 | |
| XM_005246022.3 | 3 | 647 | XP_005246079.1 | |
| XM_006710990.1 | 3 | 647 | XP_006711053.1 | |
| XM_006710991.1 | 3 | 443 | XP_006711054.1 | |
| AK027902.1 | 3 | 462 | BAB55444.1 | |
| AK074154.1 | 2 | 470 | BAB84980.1 | |
| AK096437.1 | 3 | 644 | ||
| AK097172.1 | BAC04970.1 | |||
| AK293038.1 | 2 | 758 | ||
| AY037157.1 | 3 | 419 | AAK67636.1 | |
| BC009224.2 | AAH09224.2 | |||
| BC015764.1 | 2 | 235 | AAH15764.1 | |
| BC035725.1 | 3 | 399 | AAH35725.1 | |
| DA435960.1 | 3 | 492 | ||
| DA948740.1 | 3 | 469 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526051 | Chr.1:27284255 - 27445496 on Build GRCh38 | Gain |
|
| nsv516766 | Chr.1:27335744 - 27405147 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map