Assay Details
Target Gene Details
Entrez Gene ID: | 6375 |
Gene Name: | X-C motif chemokine ligand 1 |
Gene Aliases: |
ATAC, LPTN, LTN, SCM-1, SCM-1a, SCM1, SCM1A, SCYC1 |
Location: |
Chr.1:168574128-168582077 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XCL1 | NM_002995.2 | 3 | 1275 | NP_002986.1 |
| D43768.1 | 3 | 1130 | BAA07825.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831836 | Chr.1:168569541 - 168747718 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map