Assay Details
Target Gene Details
Entrez Gene ID: | 64129 |
Gene Name: | tubulointerstitial nephritis antigen like 1 |
Gene Aliases: |
ARG1, LCN7, LIECG3, TINAGRP |
Location: |
Chr.1:31576384-31587686 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TINAGL1 | NM_001204414.1 | 6 | 823 | NP_001191343.1 |
| NM_001204415.1 | 6 | 524 | NP_001191344.1 | |
| NM_022164.2 | 7 | 916 | NP_071447.1 | |
| XM_005271106.2 | 7 | 916 | XP_005271163.1 | |
| XM_005271107.2 | 6 | 561 | XP_005271164.1 | |
| XM_011541946.1 | 5 | 628 | XP_011540248.1 | |
| AB050716.1 | 7 | 866 | BAB18636.1 | |
| AF205436.1 | 7 | 842 | AAG33699.1 | |
| AF236150.1 | 7 | 844 | AAG38876.1 | |
| AF289569.1 | 2 | 1374 | AAL55753.1 | |
| AK027839.1 | 1 | 60 | BAB55403.1 | |
| AK074124.1 | 4 | 2227 | BAB84950.1 | |
| AK075398.1 | 7 | 889 | BAC11596.1 | |
| AK292770.1 | 7 | 890 | ||
| AK298038.1 | 5 | 1018 | ||
| AK298382.1 | 6 | 793 | ||
| AK301070.1 | 7 | 926 | ||
| AY358421.1 | 7 | 889 | AAQ88787.1 | |
| BC009048.1 | 5 | 447 | AAH09048.1 | |
| BC064633.1 | 7 | 874 | AAH64633.1 | |
| BM923842.1 | 6 | 524 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011229 | Chr.1:31566071 - 31606805 on Build GRCh38 | Loss |
|
| nsv10062 | Chr.1:31571300 - 31623381 on Build GRCh38 | Gain+Loss |
|
| nsv950384 | Chr.1:31571200 - 31588799 on Build GRCh38 | Deletion |
|
| esv2758930 | Chr.1:31209533 - 32000730 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map