Assay Details
Target Gene Details
Entrez Gene ID: | 7067 |
Gene Name: | thyroid hormone receptor, alpha |
Gene Aliases: |
AR7, CHNG6, EAR7, ERB-T-1, ERBA, ERBA1, NR1A1, THRA1, THRA2, c-ERBA-1 |
Location: |
Chr.17:40062193-40093867 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THRA | NM_001190918.1 | 2 | 442 | NP_001177847.1 |
| NM_001190919.1 | 2 | 423 | NP_001177848.1 | |
| NM_003250.5 | 2 | 442 | NP_003241.2 | |
| NM_199334.3 | 2 | 442 | NP_955366.1 | |
| AB209346.1 | 2 | 492 | BAD92583.1 | |
| AK290071.1 | 2 | 269 | ||
| AK290530.1 | 2 | 423 | ||
| AK311344.1 | 2 | 217 | ||
| BC000261.1 | 2 | 291 | AAH00261.1 | |
| BC002728.2 | 2 | 291 | AAH02728.1 | |
| BC008851.2 | 2 | 513 | ||
| BC035137.1 | 2 | 451 | AAH35137.1 | |
| M24899.1 | 2 | 278 | AAA35783.1 | |
| X55005.1 | 2 | 333 | CAA38749.1 | |
| Y00479.1 | 2 | 202 | CAA68539.1 |
More Information
Set Membership: |
|

Genomic Map