Assay Details
Target Gene Details
Entrez Gene ID: | 23410 |
Gene Name: | sirtuin 3 |
Gene Aliases: |
SIR2L3 |
Location: |
Chr.11:215030-236950 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIRT3 | NM_001017524.2 | NP_001017524.1 | ||
| NM_012239.5 | NP_036371.1 | |||
| XM_005252835.1 | XP_005252892.1 | |||
| XM_011519956.1 | XP_011518258.1 | |||
| XM_011519957.1 | XP_011518259.1 | |||
| XM_017017428.1 | XP_016872917.1 | |||
| XM_017017429.1 | XP_016872918.1 | |||
| XM_017017430.1 | XP_016872919.1 | |||
| XM_017017431.1 | XP_016872920.1 | |||
| AF083108.2 | AAD40851.1 | |||
| AK074992.1 | ||||
| AK298572.1 | ||||
| AK299009.1 | ||||
| AK299438.1 | ||||
| AK301558.1 | ||||
| AK301979.1 | ||||
| AK316175.1 | ||||
| AL137276.1 | CAB70674.1 | |||
| AL535769.3 | ||||
| BC001042.2 | AAH01042.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32593 | Chr.11:218394 - 285937 on Build GRCh38 | Loss |
|
| esv3625061 | Chr.11:195766 - 233956 on Build GRCh38 | Gain |
|
| esv3625060 | Chr.11:194412 - 252166 on Build GRCh38 | Gain |
|
| nsv552765 | Chr.11:199256 - 320836 on Build GRCh38 | Gain |
|
| dgv1012n100 | Chr.11:198510 - 427836 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map