Assay Details
Target Gene Details
Entrez Gene ID: | 4254 |
Gene Name: | KIT ligand |
Gene Aliases: |
DCUA, DFNA69, FPH2, FPHH, KL-1, Kitl, MGF, SCF, SF, SHEP7 |
Location: |
Chr.12:88492793-88580473 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KITLG | NM_000899.4 | NP_000890.1 | ||
| NM_003994.5 | NP_003985.2 | |||
| AF119835.1 | AAD22048.1 | |||
| AF400436.1 | AAK92485.1 | |||
| AF400437.1 | AAK92486.1 | |||
| AK290319.1 | ||||
| AK293002.1 | ||||
| BC069733.1 | AAH69733.1 | |||
| BC069783.1 | AAH69783.1 | |||
| BC069797.1 | AAH69797.1 | |||
| BC074725.2 | AAH74725.1 | |||
| BC126166.1 | ||||
| BC143899.1 | ||||
| CR749222.1 | CAH18078.1 | |||
| M59964.1 | AAA85450.1 | |||
| S42571.1 | AAB22846.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483111 | Chr.12:86306223 - 88606223 on Build GRCh38 | Loss |
|
| nsv559704 | Chr.12:88372790 - 88645023 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map