Assay Details
Target Gene Details
Entrez Gene ID: | 54866 |
Gene Name: | protein phosphatase 1 regulatory inhibitor subunit 14D |
Gene Aliases: |
CPI17-like, GBPI-1, GBPI1 |
Location: |
Chr.15:40815445-40828793 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R14D | NM_001130143.1 | NP_001123615.1 | ||
| NM_017726.7 | NP_060196.1 | |||
| XM_017022372.1 | XP_016877861.1 | |||
| AK000258.1 | BAA91037.1 | |||
| AY050671.1 | AAL25829.1 | |||
| BC096716.1 | AAH96716.1 | |||
| BC098124.1 | AAH98124.1 | |||
| BC098150.1 | AAH98150.1 | |||
| BC098341.1 | AAH98341.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054764 | Chr.15:40776427 - 40904212 on Build GRCh38 | Gain |
|
| nsv569228 | Chr.15:40814287 - 40901953 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map