Assay Details
Target Gene Details
Entrez Gene ID: | 5598 |
Gene Name: | mitogen-activated protein kinase 7 |
Gene Aliases: |
BMK1, ERK4, ERK5, PRKM7 |
Location: |
Chr.17:19377721-19383544 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK7 | NM_002749.3 | 4 | 1331 | NP_002740.2 |
| NM_139032.2 | 2 | 1139 | NP_620601.1 | |
| NM_139033.2 | 4 | 1542 | NP_620602.2 | |
| NM_139034.2 | 4 | 1243 | NP_620603.2 | |
| XM_006721557.3 | 5 | 1307 | XP_006721620.1 | |
| XM_006721558.3 | 5 | 1560 | XP_006721621.1 | |
| XM_006721559.3 | 5 | 1469 | XP_006721622.1 | |
| XM_011523957.2 | 2 | 1046 | XP_011522259.1 | |
| AB209611.1 | 1 | 2837 | BAD92848.1 | |
| AK295376.1 | 4 | 1236 | ||
| AY534741.1 | 3 | 1169 | AAS38577.1 | |
| BC007404.2 | 2 | 1107 | AAH07404.1 | |
| BC007992.2 | 2 | 1107 | AAH07992.1 | |
| BC009963.2 | 4 | 1263 | AAH09963.1 | |
| BC030134.1 | 4 | 1178 | AAH30134.1 | |
| U25278.1 | 4 | 1236 | AAA81381.1 | |
| U29725.1 | 4 | 1377 | AAA82931.1 | |
| U29726.1 | 4 | 1210 | AAA82932.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3582561 | Chr.17:19240831 - 19585268 on Build GRCh38 | Loss |
|
| nsv1061317 | Chr.17:19047542 - 20322438 on Build GRCh38 | Gain |
|
| nsv953845 | Chr.17:19377788 - 19387787 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map