Assay Details
Target Gene Details
Entrez Gene ID: | 124637 |
Gene Name: | cytochrome b5 domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.17:7857746-7864417 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYB5D1 | NM_144607.4 | 4 | 2155 | NP_653208.2 |
| XM_005256440.3 | 4 | 1866 | XP_005256497.1 | |
| XM_011523638.2 | XP_011521940.1 | |||
| AK022811.1 | 1 | 776 | ||
| AK057061.1 | BAB71357.1 | |||
| AK125598.1 | 1 | 1749 | ||
| BC060779.1 | 4 | 1907 | AAH60779.1 |
Target Gene Details
Entrez Gene ID: | 84316 |
Gene Name: | N(alpha)-acetyltransferase 38, NatC auxiliary subunit |
Gene Aliases: |
LSMD1, PFAAP2 |
Location: |
Chr.17:7856685-7885465 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA38 | XM_011524037.2 | XP_011522339.1 | ||
| XM_017025225.1 | XP_016880714.1 | |||
| XM_017025226.1 | XP_016880715.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv510n67 | Chr.17:7852429 - 7911448 on Build GRCh38 | Gain |
|
| nsv833351 | Chr.17:7755691 - 7893863 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map