Assay Details
Target Gene Details
Entrez Gene ID: | 203074 |
Gene Name: | protease, serine 55 |
Gene Aliases: |
CT153, T-SP1, TSP1, UNQ9391 |
Location: |
Chr.8:10525331-10554166 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS55 | NM_001197020.1 | NP_001183949.1 | ||
| NM_198464.3 | NP_940866.2 | |||
| XM_005272372.3 | 1 | 255 | XP_005272429.2 | |
| XM_011543814.2 | 1 | 263 | XP_011542116.2 | |
| XM_017013182.1 | 1 | 260 | XP_016868671.1 | |
| AY358867.1 | AAQ89226.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv610268 | Chr.8:10474126 - 10629142 on Build GRCh38 | Loss |
|
| nsv1026918 | Chr.8:10502124 - 10532942 on Build GRCh38 | Loss |
|
| nsv1025557 | Chr.8:10359216 - 10697677 on Build GRCh38 | Gain |
|
| nsv1030934 | Chr.8:10207659 - 10703651 on Build GRCh38 | Gain |
|
| nsv610267 | Chr.8:10411226 - 10669232 on Build GRCh38 | Gain |
|
| esv3572508 | Chr.8:10443405 - 10588192 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map