Assay Details
Target Gene Details
Entrez Gene ID: | 144233 |
Gene Name: | BCDIN3 domain containing RNA methyltransferase |
Gene Aliases: |
- |
Location: |
Chr.12:49836043-49843129 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCDIN3D | NM_181708.2 | 1 | 213 | NP_859059.1 |
| AK292194.1 | 1 | 190 | ||
| BC053560.1 | 1 | 176 | AAH53560.1 | |
| CD690663.1 | 1 | 235 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522972 | Chr.12:49802611 - 50017048 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map