Assay Details
Target Gene Details
Entrez Gene ID: | 54814 |
Gene Name: | glutaminyl-peptide cyclotransferase like |
Gene Aliases: |
gQC |
Location: |
Chr.19:45692402-45703990 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QPCTL | NM_001163377.1 | 1 | 383 | NP_001156849.1 |
| NM_017659.3 | 1 | 383 | NP_060129.2 | |
| XM_011527048.2 | 1 | 464 | XP_011525350.1 | |
| XM_017026900.1 | 1 | 462 | XP_016882389.1 | |
| AK000091.1 | 1 | 192 | BAA90938.1 | |
| AK172764.1 | 1 | 191 | BAD18747.1 | |
| AK222636.1 | 1 | 176 | BAD96356.1 | |
| BC011553.1 | 1 | 169 | AAH11553.1 | |
| BP216511.1 | 1 | 383 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644509 | Chr.19:45677013 - 45696301 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map