Assay Details
Target Gene Details
Entrez Gene ID: | 8536 |
Gene Name: | calcium/calmodulin dependent protein kinase I |
Gene Aliases: |
CAMKI |
Location: |
Chr.3:9757345-9769987 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAMK1 | NM_003656.4 | NP_003647.1 | ||
| XM_005265516.1 | XP_005265573.1 | |||
| XM_005265517.2 | XP_005265574.1 | |||
| XM_017007354.1 | XP_016862843.1 | |||
| AA724193.1 | ||||
| AB209395.1 | BAD92632.1 | |||
| L41816.1 | AAA99458.1 |
Target Gene Details
Entrez Gene ID: | 4968 |
Gene Name: | 8-oxoguanine DNA glycosylase |
Gene Aliases: |
HMMH, HOGG1, MUTM, OGH1 |
Location: |
Chr.3:9749944-9788246 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OGG1 | NM_002542.5 | 7 | 1559 | NP_002533.1 |
| NM_016819.3 | 6 | 1803 | NP_058212.1 | |
| NM_016820.3 | 7 | 1576 | NP_058213.1 | |
| NM_016821.2 | NP_058214.1 | |||
| NM_016826.2 | NP_058434.1 | |||
| NM_016827.2 | NP_058436.1 | |||
| NM_016828.2 | NP_058437.1 | |||
| NM_016829.2 | NP_058438.1 | |||
| XM_011533760.1 | XP_011532062.1 | |||
| XM_017006493.1 | 6 | 1403 | XP_016861982.1 | |
| XM_017006494.1 | XP_016861983.1 | |||
| XM_017006495.1 | 6 | 1420 | XP_016861984.1 | |
| XM_017006496.1 | XP_016861985.1 | |||
| XM_017006497.1 | XP_016861986.1 | |||
| XM_017006498.1 | XP_016861987.1 | |||
| XM_017006499.1 | XP_016861988.1 | |||
| AB000410.1 | 7 | 1484 | BAA19103.1 | |
| AB019528.1 | BAA76635.1 | |||
| AB019529.1 | BAA76636.1 | |||
| AB019530.1 | BAA76637.1 | |||
| AB019531.1 | BAA76638.1 | |||
| AB019532.1 | 4 | 1059 | BAA76639.1 | |
| AB037879.1 | BAB13294.1 | |||
| AF003595.1 | 7 | 1272 | AAB61340.1 | |
| AF026691.1 | 7 | 1440 | AAB84013.1 | |
| AK289858.1 | 7 | 1512 | ||
| BC000657.2 | 7 | 1513 | AAH00657.1 | |
| U88527.1 | 7 | 1526 | AAB68614.1 | |
| U88620.1 | AAB68615.1 | |||
| U96710.1 | 7 | 1290 | AAB81132.1 | |
| Y11731.2 | 7 | 1524 | CAA72414.1 | |
| Y11838.1 | 7 | 1523 | CAA72536.1 | |
| Y13277.1 | CAA73726.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010768 | Chr.3:9502870 - 9873902 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map