Assay Details
Target Gene Details
Entrez Gene ID: | 140730 |
Gene Name: | regulating synaptic membrane exocytosis 4 |
Gene Aliases: |
C20orf190, RIM 4, RIM-4, RIM4, RIM4-gamma, RIM4gamma |
Location: |
Chr.20:44751804-44810338 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIMS4 | NM_001205317.1 | 6 | 2916 | NP_001192246.1 |
| NM_182970.3 | 6 | 2913 | NP_892015.1 | |
| AK055698.1 | 1 | 446 | ||
| AK095716.1 | ||||
| AK124626.1 | 2 | 2307 | ||
| AK126807.1 | 6 | 2816 | ||
| BX648947.1 | 5 | 2686 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv586076 | Chr.20:44706933 - 44779789 on Build GRCh38 | Gain |
|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map