Assay Details
Target Gene Details
Entrez Gene ID: | 220001 |
Gene Name: | von Willebrand factor C and EGF domains |
Gene Aliases: |
URG11, VWC1 |
Location: |
Chr.11:61258286-61295316 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VWCE | NM_152718.2 | 20 | 3061 | NP_689931.2 |
| XM_006718460.1 | 20 | 3037 | XP_006718523.1 | |
| XM_006718462.1 | 18 | 2881 | XP_006718525.1 | |
| XM_011544817.1 | 17 | 2767 | XP_011543119.1 | |
| XM_017017340.1 | 20 | 2998 | XP_016872829.1 | |
| XM_017017341.1 | 19 | 2932 | XP_016872830.1 | |
| XM_017017342.1 | 19 | 2654 | XP_016872831.1 | |
| XM_017017343.1 | 21 | 2845 | XP_016872832.1 | |
| AK056571.1 | 20 | 2819 | BAB71219.1 | |
| AK302241.1 | 9 | 1300 | ||
| AY205235.1 | 18 | 2566 | AAO47606.1 | |
| BC142624.1 | 20 | 2808 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759831 | Chr.11:61147888 - 61313185 on Build GRCh38 | Gain+Loss |
|
| nsv428259 | Chr.11:61049513 - 61356164 on Build GRCh38 | Gain+Loss |
|
| esv28446 | Chr.11:61257988 - 61267462 on Build GRCh38 | Loss |
|
| nsv341 | Chr.11:61240546 - 61259664 on Build GRCh38 | Insertion |
|
| esv3626577 | Chr.11:61257434 - 61267693 on Build GRCh38 | Loss |
|
| nsv832177 | Chr.11:61185118 - 61356164 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map