Assay Details
Target Gene Details
Entrez Gene ID: | 151393 |
Gene Name: | regulator of microtubule dynamics 2 |
Gene Aliases: |
BLOCK18, FAM82A, FAM82A1, PRO34163, PYST9371, RMD-2, RMD2, RMD4 |
Location: |
Chr.2:37920789-38068032 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Target Gene Details
Entrez Gene ID: | 101410544 |
Gene Name: | RMDN2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:37950334-38036341 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RMDN2-AS1 | NR_102712.1 | |||
| DB142860.1 |
More Information
Set Membership: |
|

Genomic Map