Assay Details
Target Gene Details
Entrez Gene ID: | 55176 |
Gene Name: | Sec61 translocon alpha 2 subunit |
Gene Aliases: |
- |
Location: |
Chr.10:12129641-12169958 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC61A2 | NM_001142627.2 | 8 | 823 | NP_001136099.1 |
| NM_001142628.1 | 7 | 757 | NP_001136100.1 | |
| NM_018144.3 | 8 | 823 | NP_060614.2 | |
| NR_024576.2 | 8 | 823 | ||
| NR_024577.2 | 9 | 956 | ||
| AF346603.1 | 8 | 764 | AAK29084.1 | |
| AK001440.1 | 1 | 58 | BAA91692.1 | |
| AK022027.1 | 2 | 206 | BAB13955.1 | |
| AK022640.1 | 8 | 758 | BAB14148.1 | |
| AK057532.1 | 9 | 897 | ||
| AK292295.1 | 8 | 773 | ||
| AK301841.1 | 7 | 710 | ||
| BC026179.1 | 8 | 773 | AAH26179.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035709 | Chr.10:12054410 - 12183299 on Build GRCh38 | Gain |
|
| nsv5820 | Chr.10:12155307 - 12187443 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map