Assay Details
Target Gene Details
Entrez Gene ID: | 9046 |
Gene Name: | docking protein 2 |
Gene Aliases: |
p56DOK, p56dok-2 |
Location: |
Chr.8:21908873-21913717 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOK2 | NM_001317800.1 | NP_001304729.1 | ||
| NM_003974.3 | NP_003965.2 | |||
| NM_201349.2 | NP_958728.1 | |||
| XM_005273680.4 | XP_005273737.1 | |||
| AF034970.1 | AAC13265.1 | |||
| AK226110.1 | ||||
| AK292126.1 | ||||
| AK296671.1 | ||||
| BC032623.1 | AAH32623.1 | |||
| CB270269.1 | ||||
| DC295259.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759604 | Chr.8:20675533 - 23254096 on Build GRCh38 | Loss |
|
| dgv7131n100 | Chr.8:21905882 - 21974708 on Build GRCh38 | Gain |
|
| nsv1023009 | Chr.8:21908590 - 21952074 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map