Assay Details
Target Gene Details
Entrez Gene ID: | 9214 |
Gene Name: | Fc fragment of IgM receptor |
Gene Aliases: |
FAIM3, TOSO |
Location: |
Chr.1:206903282-206922033 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCMR | NM_001142473.1 | NP_001135945.1 | ||
| NM_001193338.1 | 2 | 255 | NP_001180267.1 | |
| NM_005449.4 | 2 | 398 | NP_005440.1 | |
| XM_005273351.3 | 2 | 398 | XP_005273408.1 | |
| XM_005273352.4 | 2 | 375 | XP_005273409.1 | |
| AF057557.1 | 2 | 238 | AAC18830.1 | |
| AK292007.1 | 2 | 306 | ||
| AK297053.1 | 2 | 306 | ||
| AK301187.1 | ||||
| AK316336.1 | ||||
| BC006401.2 | 2 | 363 | AAH06401.1 | |
| BP225124.1 | ||||
| BP305798.1 | ||||
| BT006797.1 | 2 | 219 | AAP35443.1 | |
| DA971987.1 | 2 | 398 | ||
| DB122623.1 | 3 | 356 | ||
| DC393681.1 | ||||
| GQ160900.1 | 2 | 264 | ||
| GQ160901.1 | 2 | 280 | ||
| HM480394.1 | 2 | 232 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010486 | Chr.1:206876486 - 207031685 on Build GRCh38 | Gain |
|
| nsv468049 | Chr.1:206894495 - 206963993 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map