Assay Details
Target Gene Details
Entrez Gene ID: | 9344 |
Gene Name: | TAO kinase 2 |
Gene Aliases: |
MAP3K17, PSK, PSK1, PSK1-BETA, TAO1, TAO2 |
Location: |
Chr.16:29973641-29992261 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAOK2 | NM_001252043.1 | NP_001238972.1 | ||
| NM_004783.3 | NP_004774.1 | |||
| NM_016151.3 | NP_057235.2 | |||
| XM_011545982.1 | XP_011544284.1 | |||
| XM_011545983.2 | XP_011544285.1 | |||
| XM_011545984.1 | XP_011544286.1 | |||
| XM_011545985.1 | XP_011544287.1 | |||
| XM_011545986.2 | XP_011544288.1 | |||
| AB020688.1 | ||||
| AF061943.1 | AAD45616.1 | |||
| AF263313.1 | AAG38503.1 | |||
| AK291473.1 | ||||
| BC015204.1 | ||||
| BC031825.2 | AAH31825.1 | |||
| BC051798.1 | AAH51798.1 | |||
| BC136653.1 | ||||
| BC136655.1 | ||||
| BC142663.1 | ||||
| BC144344.1 | ||||
| BC151221.1 | ||||
| BC152413.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638378 | Chr.16:29973809 - 30006402 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map