Assay Details
Target Gene Details
Entrez Gene ID: | 55749 |
Gene Name: | cell division cycle and apoptosis regulator 1 |
Gene Aliases: |
- |
Location: |
Chr.10:68721144-68792377 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCAR1 | NM_001282959.1 | NP_001269888.1 | ||
| NM_001282960.1 | NP_001269889.1 | |||
| NM_018237.3 | NP_060707.2 | |||
| NR_104262.1 | ||||
| AF465616.1 | AAO17319.1 | |||
| AK001701.1 | BAA91847.1 | |||
| AK023438.1 | BAB14574.1 | |||
| AK122871.1 | ||||
| AK292649.1 | ||||
| AK298004.1 | ||||
| AK299307.1 | ||||
| AK307550.1 | ||||
| AK307671.1 | ||||
| AK307704.1 | ||||
| AY249140.1 | AAP82002.1 | |||
| BC108682.1 | AAI08683.1 | |||
| BC130626.1 | ||||
| BC132725.1 | ||||
| BM786237.1 | ||||
| BX647943.1 | ||||
| DA511293.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052569 | Chr.10:68650779 - 68781892 on Build GRCh38 | Gain |
|
| nsv551435 | Chr.10:68642219 - 68796440 on Build GRCh38 | Loss |
|
| nsv6999 | Chr.10:68719566 - 68732782 on Build GRCh38 | Insertion |
|
| dgv919n100 | Chr.10:68400084 - 68752800 on Build GRCh38 | Gain |
|
| esv2759764 | Chr.10:68697962 - 68871556 on Build GRCh38 | Loss |
|
| nsv1047983 | Chr.10:68414663 - 68849311 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map