Assay Details
Target Gene Details
Entrez Gene ID: | 22953 |
Gene Name: | purinergic receptor P2X 2 |
Gene Aliases: |
DFNA41, P2X2 |
Location: |
Chr.12:132618780-132623678 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2RX2 | NM_001282164.1 | 11 | 1402 | NP_001269093.1 |
| NM_001282165.1 | 10 | 1802 | NP_001269094.1 | |
| NM_012226.4 | 9 | 1489 | NP_036358.2 | |
| NM_016318.3 | 10 | 1633 | NP_057402.1 | |
| NM_170682.3 | 11 | 1705 | NP_733782.1 | |
| NM_170683.3 | 10 | 1783 | NP_733783.1 | |
| NM_174872.2 | 9 | 1429 | NP_777361.1 | |
| NM_174873.2 | 12 | 1504 | NP_777362.1 | |
| XM_005266154.3 | 10 | 1812 | XP_005266211.1 | |
| XM_005266155.4 | 11 | 1474 | XP_005266212.1 | |
| XM_005266156.4 | 11 | 1576 | XP_005266213.1 | |
| XM_011534786.2 | 9 | 1695 | XP_011533088.1 | |
| XM_017019035.1 | 12 | 1396 | XP_016874524.1 | |
| AF190822.1 | 11 | 1668 | AAF19170.1 | |
| BC109200.2 | 10 | 1765 | AAI09201.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952512 | Chr.12:132192256 - 132711214 on Build GRCh38 | Deletion |
|
| nsv561109 | Chr.12:132618549 - 132626173 on Build GRCh38 | Loss |
|
| esv2422284 | Chr.12:132604248 - 132708921 on Build GRCh38 | Deletion |
|
| nsv826563 | Chr.12:132125383 - 132622340 on Build GRCh38 | Gain |
|
| nsv1037074 | Chr.12:132613063 - 132801953 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map