Assay Details
Target Gene Details
Entrez Gene ID: | 2941 |
Gene Name: | glutathione S-transferase alpha 4 |
Gene Aliases: |
GSTA4-4, GTA4 |
Location: |
Chr.6:52977948-52995380 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSTA4 | NM_001512.3 | 7 | 1113 | NP_001503.1 |
| XM_005249035.3 | 7 | 1206 | XP_005249092.1 | |
| XM_011514534.2 | 6 | 1056 | XP_011512836.1 | |
| XM_011514535.2 | 6 | 1152 | XP_011512837.1 | |
| AA709059.1 | 1 | 240 | ||
| AF020918.1 | 7 | 1033 | AAD04711.1 | |
| AF025887.1 | 7 | 1036 | AAC39695.1 | |
| AK054804.1 | 5 | 1470 | ||
| BC015523.2 | 6 | 983 | AAH15523.1 | |
| BC063439.1 | 5 | 950 | AAH63439.1 | |
| CR749474.1 | 3 | 4412 | CAH18304.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759430 | Chr.6:52928056 - 52996406 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map