Assay Details
Target Gene Details
Entrez Gene ID: | 1586 |
Gene Name: | cytochrome P450 family 17 subfamily A member 1 |
Gene Aliases: |
CPT7, CYP17, P450C17, S17AH |
Location: |
Chr.10:102830531-102837533 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP17A1 | NM_000102.3 | NP_000093.1 | ||
| AK094106.1 | ||||
| AK289898.1 | ||||
| AK308466.1 | ||||
| AK312235.1 | ||||
| BC062997.1 | AAH62997.1 | |||
| BC063388.1 | AAH63388.1 | |||
| BT020000.1 | AAV38803.1 | |||
| M14564.1 | AAA52151.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7527 | Chr.10:102805716 - 102839530 on Build GRCh38 | Insertion |
|
| nsv526666 | Chr.10:102824573 - 102837723 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map