Assay Details
Target Gene Details
Entrez Gene ID: | 64943 |
Gene Name: | 5'-nucleotidase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.3:52524369-52535077 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NT5DC2 | NM_001134231.1 | NP_001127703.1 | ||
| NM_022908.2 | NP_075059.1 | |||
| XM_005265406.2 | XP_005265463.1 | |||
| XM_006713303.3 | XP_006713366.1 | |||
| XM_006713305.2 | XP_006713368.1 | |||
| AF131781.1 | AAD20044.1 | |||
| AK022504.1 | BAB14064.1 | |||
| AK023995.1 | BAB14763.1 | |||
| BC014550.1 | AAH14550.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834697 | Chr.3:52366562 - 52545401 on Build GRCh38 | Loss |
|
| nsv954862 | Chr.3:52476885 - 52530884 on Build GRCh38 | Deletion |
|
| dgv8326n54 | Chr.3:52501570 - 52538040 on Build GRCh38 | Loss |
|
| nsv3820 | Chr.3:52496531 - 52530448 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map