Assay Details
Target Gene Details
Entrez Gene ID: | 54822 |
Gene Name: | transient receptor potential cation channel subfamily M member 7 |
Gene Aliases: |
ALSPDC, CHAK, CHAK1, LTRPC7, LTrpC-7, TRP-PLIK |
Location: |
Chr.15:50557158-50686835 on Build GRCh38 |
Assay Gene Location: | Within Exon 40 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPM7 | NM_001301212.1 | 36 | 5536 | NP_001288141.1 |
| NM_017672.5 | 36 | 5539 | NP_060142.3 | |
| XM_005254486.3 | 37 | 5594 | XP_005254543.1 | |
| XM_017022350.1 | 37 | 5359 | XP_016877839.1 | |
| XM_017022351.1 | 37 | 5356 | XP_016877840.1 | |
| XM_017022352.1 | 36 | 5299 | XP_016877841.1 | |
| XM_017022353.1 | 36 | 5296 | XP_016877842.1 | |
| XM_017022354.1 | 37 | 5424 | XP_016877843.1 | |
| XM_017022355.1 | 35 | 5226 | XP_016877844.1 | |
| AF346629.2 | 36 | 5492 | AAK19738.2 | |
| AK075193.1 | 2 | 244 | BAC11462.1 | |
| AK098584.1 | 7 | 814 | ||
| AK172800.1 | 13 | 1984 | BAD18773.1 | |
| AY032950.1 | 36 | 5528 | AAK44211.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055119 | Chr.15:50569273 - 50608488 on Build GRCh38 | Gain |
|
| dgv317n27 | Chr.15:50478671 - 50897510 on Build GRCh38 | Gain |
|
| nsv1050385 | Chr.15:50551298 - 50657601 on Build GRCh38 | Gain |
|
| nsv524613 | Chr.15:50364252 - 50576582 on Build GRCh38 | Loss |
|
| nsv1047443 | Chr.15:50452584 - 51354826 on Build GRCh38 | Gain |
|
| nsv569398 | Chr.15:50496928 - 51002798 on Build GRCh38 | Loss |
|
| dgv2594n100 | Chr.15:50469213 - 50823338 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map