Assay Details
Target Gene Details
Entrez Gene ID: | 102465535 |
Gene Name: | microRNA 6888 |
Gene Aliases: |
hsa-mir-6888 |
Location: |
Chr.2:159186835-159186901 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR6888 | NR_106948.1 |
Target Gene Details
Entrez Gene ID: | 85461 |
Gene Name: | tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1 |
Gene Aliases: |
ROLSB, TANC |
Location: |
Chr.2:158968600-159232659 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 23 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011598 | Chr.2:159175496 - 159224825 on Build GRCh38 | Gain |
|
| nsv1010774 | Chr.2:159012247 - 160175703 on Build GRCh38 | Gain |
|
| dgv4096n100 | Chr.2:159097908 - 159201276 on Build GRCh38 | Gain |
|
| esv3593110 | Chr.2:159183805 - 159201028 on Build GRCh38 | Loss |
|
| nsv1008665 | Chr.2:159163999 - 159322188 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map