Assay Details
Target Gene Details
Entrez Gene ID: | 79901 |
Gene Name: | cytochrome b reductase 1 |
Gene Aliases: |
CYB561A2, DCYTB, FRRS3 |
Location: |
Chr.2:171522247-171558133 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYBRD1 | NM_001127383.1 | NP_001120855.1 | ||
| NM_001256909.1 | 2 | 130 | NP_001243838.1 | |
| NM_024843.3 | 2 | 468 | NP_079119.3 | |
| AK027115.1 | 2 | 310 | BAB15661.1 | |
| AK128495.1 | BAC87466.1 | |||
| AK225459.1 | 2 | 310 | ||
| AK291731.1 | 2 | 318 | ||
| AK301485.1 | 2 | 130 | ||
| AK316588.1 | 2 | 347 | ||
| AL136693.1 | 2 | 351 | CAB66628.1 | |
| BC065290.1 | 2 | 303 | AAH65290.1 | |
| CB266594.1 | ||||
| CX866825.1 | 2 | 359 | ||
| DB235339.1 | 2 | 468 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160884 | Chr.2:171536097 - 171553747 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map