Assay Details
Target Gene Details
Entrez Gene ID: | 9501 |
Gene Name: | rabphilin 3A-like (without C2 domains) |
Gene Aliases: |
NOC2 |
Location: |
Chr.17:212389-386254 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPH3AL | NM_001190411.1 | NP_001177340.1 | ||
| NM_001190412.1 | NP_001177341.1 | |||
| NM_001190413.1 | NP_001177342.1 | |||
| NM_006987.3 | NP_008918.1 | |||
| XM_011535364.2 | XP_011533666.1 | |||
| XM_011535365.2 | XP_011533667.1 | |||
| XM_011535366.2 | XP_011533668.1 | |||
| XM_011535367.2 | XP_011533669.1 | |||
| XM_011535368.2 | XP_011533670.1 | |||
| XM_011535369.2 | XP_011533671.1 | |||
| XM_017025367.1 | XP_016880856.1 | |||
| XM_017025368.1 | XP_016880857.1 | |||
| XM_017025369.1 | XP_016880858.1 | |||
| XM_017025370.1 | XP_016880859.1 | |||
| AF129812.1 | AAD45582.1 | |||
| AK000469.1 | BAA91186.1 | |||
| AK291950.1 | ||||
| BC005153.1 | AAH05153.1 | |||
| BC093776.1 | AAH93776.1 | |||
| BC113413.1 | ||||
| BU197069.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892942 | Chr.17:242355 - 253678 on Build GRCh38 | Loss |
|
| nsv952087 | Chr.17:244610 - 267509 on Build GRCh38 | Deletion |
|
| nsv1065720 | Chr.17:239022 - 264687 on Build GRCh38 | Loss |
|
| nsv469538 | Chr.17:150208 - 266971 on Build GRCh38 | Loss |
|
| esv3639615 | Chr.17:177867 - 302096 on Build GRCh38 | Gain |
|
| nsv1056138 | Chr.17:150721 - 252418 on Build GRCh38 | Loss |
|
| nsv1937 | Chr.17:236742 - 274809 on Build GRCh38 | Insertion |
|
| nsv509642 | Chr.17:191012 - 394464 on Build GRCh38 | Insertion |
|
| nsv573933 | Chr.17:191293 - 286907 on Build GRCh38 | Gain |
|
| nsv573947 | Chr.17:201297 - 421142 on Build GRCh38 | Gain |
|
| esv2422420 | Chr.17:210752 - 1113141 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map