Assay Details
Target Gene Details
Entrez Gene ID: | 84881 |
Gene Name: | RNA pseudouridylate synthase domain containing 4 |
Gene Aliases: |
- |
Location: |
Chr.11:126202094-126211692 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPUSD4 | NM_001144827.1 | NP_001138299.1 | ||
| NM_032795.2 | NP_116184.2 | |||
| XM_011543039.2 | XP_011541341.1 | |||
| AK027400.1 | BAB55086.1 | |||
| AK225320.1 | ||||
| AK308059.1 | ||||
| BC014131.2 | AAH14131.1 | |||
| DB051034.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556529 | Chr.11:126204297 - 126264724 on Build GRCh38 | Loss |
|
| nsv556527 | Chr.11:126172311 - 126211508 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs141455937] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map