Assay Details
Target Gene Details
Entrez Gene ID: | 23351 |
Gene Name: | KH and NYN domain containing |
Gene Aliases: |
KIAA0323 |
Location: |
Chr.14:24423186-24441342 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KHNYN | NM_001290256.1 | 8 | 4663 | NP_001277185.1 |
| NM_001290257.1 | 8 | 4598 | NP_001277186.1 | |
| NM_015299.2 | 8 | 4726 | NP_056114.1 | |
| XM_005267474.4 | 8 | 4682 | XP_005267531.1 | |
| XM_011536590.2 | 8 | 4832 | XP_011534892.1 | |
| AB002321.1 | 8 | 4663 |
Target Gene Details
Entrez Gene ID: | 56948 |
Gene Name: | short chain dehydrogenase/reductase family 39U member 1 |
Gene Aliases: |
C14orf124, HCDI |
Location: |
Chr.14:24439766-24442905 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDR39U1 | NM_001290292.1 | NP_001277221.1 | ||
| NM_001290293.2 | NP_001277222.1 | |||
| NM_001308075.1 | NP_001295004.1 | |||
| NM_020195.2 | NP_064580.2 | |||
| XM_011536975.1 | XP_011535277.1 | |||
| XM_011536976.1 | XP_011535278.1 | |||
| AF226050.1 | AAF86950.1 | |||
| AK123513.1 | ||||
| BC000989.2 | AAH00989.1 | |||
| BX161384.1 | CAD61875.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1854n100 | Chr.14:24438841 - 24524460 on Build GRCh38 | Gain |
|
| nsv482171 | Chr.14:24439768 - 24442801 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map