Assay Details
Target Gene Details
Entrez Gene ID: | 23071 |
Gene Name: | endoplasmic reticulum protein 44 |
Gene Aliases: |
PDIA10, TXNDC4 |
Location: |
Chr.9:99979179-100099052 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERP44 | NM_015051.2 | 12 | 2005 | NP_055866.1 |
| AB011145.1 | 12 | 1929 | ||
| BX648519.1 | 14 | 2242 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050510 | Chr.9:99922036 - 99986376 on Build GRCh38 | Gain |
|
| nsv831668 | Chr.9:99805887 - 99985011 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map