Assay Details
Target Gene Details
Entrez Gene ID: | 100616211 |
Gene Name: | microRNA 4709 |
Gene Aliases: |
mir-4709 |
Location: |
Chr.14:74480133-74480204 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR4709 | NR_039858.1 |
Target Gene Details
Entrez Gene ID: | 10577 |
Gene Name: | NPC intracellular cholesterol transporter 2 |
Gene Aliases: |
EDDM1, HE1 |
Location: |
Chr.14:74479940-74493475 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPC2 | NM_006432.3 | 5 | 686 | NP_006423.1 |
| XM_017020930.1 | 4 | 1193 | XP_016876419.1 | |
| AK222474.1 | 5 | 606 | BAD96194.1 | |
| AK298975.1 | 4 | 508 | ||
| AK300879.1 | ||||
| BC002532.2 | 5 | 685 | AAH02532.1 | |
| X67698.1 | 5 | 589 | CAA47928.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3634943 | Chr.14:74426684 - 74537774 on Build GRCh38 | Gain |
|
| nsv832829 | Chr.14:74377797 - 74545100 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map